missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
This antibody is specific to DDB1.
This gene encodes the large subunit of DNA damage-binding protein which is a heterodimer composed of a large and a small subunit. This protein functions in nucleotide-excision repair. Its defective activity causes the repair defect in the patients with xeroderma pigmentosum complementation group E (XPE). However, it remains for mutation analysis to demonstrate whether the defect in XPE patients is in this gene or the gene encoding the small subunit. In addition, Best vitelliform mascular dystrophy is mapped to the same region as this gene on 11q, but no sequence alternations of this gene are demonstrated in Best disease patients.
Especificaciones
Especificaciones
| Antígeno | DDB1 |
| Aplicaciones | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 4F6C6 |
| Concentración | 0.43 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | DDB1 |
| N.º de referencia del gen | Q16531, Q3U1J4, Q9ESW0 |
| Alias de gen | DDB p127 subunit, DDB1, DDBA, DNA damage binding protein 1, DNA damage binding protein a, HBV X associated protein 1, UV damaged DNA binding factor, UV DDB 1, UV DDB1, XAP 1, XAP1, XPCE, XPE, XPE BF, XPE binding factor |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?