missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
DC-SIGN/CD209 Monoclonal antibody specifically detects DC-SIGN/CD209 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Especificaciones
Especificaciones
| Antígeno | DC-SIGN/CD209 |
| Aplicaciones | Flow Cytometry, CyTOF |
| Clasificación | Monoclonal |
| Clon | MMD3R |
| Conjugado | HRP |
| Dilución | Flow Cytometry, CyTOF-ready |
| Formulación | PBS |
| Alias de gen | CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 |
| Especie del huésped | Mouse |
| Inmunógeno | Purified mouse DC-SIGN/CD209 extracellular domain, Accession # FJ168685 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?