missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Cytosolic Sulfotransferase 1E1/SULT1E1 Antibody (SULT1E1/7124) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30064840 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
30064840 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 30064840

Brand: Novus Biologicals™ NBP327015JF549

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cytosolic Sulfotransferase 1E1/SULT1E1 Monoclonal antibody specifically detects Cytosolic Sulfotransferase 1E1/SULT1E1 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Cytosolic Sulfotransferase 1E1/SULT1E1
Aplicaciones Western Blot, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon SULT1E1/7124
Conjugado Janelia Fluor 549
Formulación 50 mM Sodium Borate
Alias de gen EC 2.8.2, EST-1, ESTMGC34459, estrone sulfotransferase, ST1E1, STEestrogen sulfotransferase, Sulfotransferase 1E1, sulfotransferase family 1E, estrogen-preferring, member 1, Sulfotransferase, estrogen-preferringEC 2.8.2.4
Especie del huésped Mouse
Inmunógeno Recombinant fragment (around aa100-294) of human Cytosolic Sulfotransferase 1E1/SULT1E1 protein (exact sequence is proprietary)
Método de purificación Protein A or G purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Breast Cancer, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 6783
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG2 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.