missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 18 Antibody (DA7), Novus Biologicals™

Código de producto. 18122204 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mg
20 ug
Tamaño de la unidad:
0.10 miligramo
20 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18122204 0.1 mg 0.10 miligramo
18475971 20 ug 20 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18122204 Proveedor Novus Biologicals N.º de proveedor NBP229409

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Cytokeratin 18 Monoclonal specifically detects Cytokeratin 18 in Human, Bovine (Negative), Canine (Negative), Hamster (Negative), Mouse (Negative), Porcine (Negative), Rat (Negative), Sheep (Negative) samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Cytokeratin 18
Aplicaciones Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon DA7
Concentración 0.2 mg/ml
Conjugado Unconjugated
Dilución Western Blot 1-3 μg/mL, Simple Western 10 μg/mL, Flow Cytometry 0.5-1 μg/million cells, ELISA 1-5 μg/mL for coating, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 μg/mL, Immunoprecipitation 1-2 μg/500 μg protein lysate, Immunohistochemistry-Paraffin 0.5-1.0 μg/mL
N.º de referencia del gen P05783
Alias de gen Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18
Símbolos de los genes KRT18
Especie del huésped Mouse
Inmunógeno Human breast cancer PMC 42 cells were used as immunogen to generate the Cytokeratin 18 (CK18) antibody.
Método de purificación Protein A or G purified
Cantidad 0.1 mg
Estado normativo RUO
Disciplina de investigación Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Phospho Specific
Primario o secundario Primary
ID de gen (Entrez) 3875
Especificidad de la prueba This monoclonal antibody reacts with a wide variety of simple epithelia. It does not react with stratified squamous epithelia. It reacts with epithelial tumors of the gastrointestinal tract, lung, breast, pancreas, ovary, and thyroid. Cytokeratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. It was reported that tissues from gastrointestinal tract are positive for both cytokeratin 8 and 18 but do not contain cytokeratin 14. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18.
Especies diana Human, Bovine (Negative), Canine (Negative), Hamster (Negative), Mouse (Negative), Porcine (Negative), Rat (Negative)
Contenido y almacenamiento Store at 4C.
Formulario Purified
Isotype IgG1 κ
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.