missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cytokeratin 10 Antibody (KRT10/6961) [DyLight 594], Novus Biologicals™
Tienda Bio Techne ProductosDescripción
Cytokeratin 10 Monoclonal antibody specifically detects Cytokeratin 10 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | Cytokeratin 10 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | KRT10/6961 |
| Conjugado | DyLight 594 |
| Formulación | 50 mM Sodium Borate |
| Alias de gen | BCIE, BIE, CK10, CK-10, cytokeratin 10, Cytokeratin-10, EHK, K10keratosis palmaris et plantaris, keratin 10, Keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris), keratin, type I cytoskeletal 10, keratin-10, KPP |
| Especie del huésped | Mouse |
| Inmunógeno | Recombinant full-length human Cytokeratin 10 epsilon protein. |
| Método de purificación | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?