missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Cyclin D1 Monoclonal antibody specifically detects Cyclin D1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Especificaciones
Especificaciones
| Antígeno | Cyclin D1 |
| Aplicaciones | Immunocytochemistry/Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | CL12328 |
| Conjugado | Alexa Fluor 405 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31 |
| Especie del huésped | Mouse |
| Inmunógeno | This antibody was developed using a synthetic peptide derived from P24385, with the exact immunogen sequence remaining proprietary. |
| Método de purificación | Protein A purified |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?