Learn More
Cyclin B1 Antibody (CCNB1/9242R), Novus Biologicals™
Código de producto. 18611804 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
20 μg
Tamaño de la unidad:
100 microgramos
20 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18611804 20 μg 20 microgramos
18654164 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18611804 Proveedor Novus Biologicals N.º de proveedor NBP32391420ug
Rabbit Monoclonal Antibody
Cyclin B1 Monoclonal antibody specifically detects Cyclin B1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Cyclin B1
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon CCNB1/9242R
Concentración 0.2 mg/mL
Conjugado Unconjugated
Dilución Immunohistochemistry-Paraffin : 1-2 μg/mL
Formulación 10 mM PBS with 0.05% BSA
Alias de gen CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1
Especie del huésped Rabbit
Inmunógeno His-tagged recombinant hamster Cyclin B1 protein
Método de purificación Protein A or G purified
Cantidad 20 μg
Estado normativo RUO
Disciplina de investigación Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cell Cycle and Replication, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Mitotic Regulators, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Stem Cell Lines, Stem Cell Markers, Tumor Suppressors
Primario o secundario Primary
ID de gen (Entrez) 891
Especies diana Human
Contenido y almacenamiento Store at 4°C.
Formulario Purified
Isotype IgG κ