missing translation for 'onlineSavingsMsg'
Learn More

Cyclin B1 Antibody (2061D), mFluor Violet 610 SE, Novus Biologicals™

Código de producto. 18591285 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18591285 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18591285 Proveedor Novus Biologicals N.º de proveedor FAB60001MFV610

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. View return policy

Rabbit Monoclonal Antibody

Cyclin B1 Monoclonal antibody specifically detects Cyclin B1 in Human samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Cyclin B1
Aplicaciones Western Blot, Immunohistochemistry, Flow Cytometry
Clasificación Monoclonal
Clon 2061D
Conjugado mFluor Violet 610 SE
Dilución Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry
Formulación 50mM Sodium Borate
Alias de gen CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1
Especie del huésped Rabbit
Inmunógeno E. coli-derived recombinant human Cyclin B1, Met1-Pro91, Accession # P14635
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cell Cycle and Replication, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Mitotic Regulators, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Stem Cell Lines, Stem Cell Markers, Tumor Suppressors
Primario o secundario Primary
ID de gen (Entrez) 891
Especies diana Human
Contenido y almacenamiento Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.