missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cyclin A2 Monoclonal antibody specifically detects Cyclin A2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antígeno | Cyclin A2 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | CCNA2/2333 |
| Conjugado | DyLight 680 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | CCN1CCNAcyclin-A, cyclin A2, Cyclin-A, cyclin-A2 |
| Especie del huésped | Mouse |
| Inmunógeno | Full length recombinant Cyclin A2 protein (Uniprot: P20248) |
| Método de purificación | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?