Learn More
CYB5B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15469-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CYB5B gene ontology annotations related to this gene include enzyme activator activity; heme binding; metal ion binding.Especificaciones
| CYB5B | |
| Polyclonal | |
| Unconjugated | |
| CYB5B | |
| CYB5 M, CYB5B, CYB5M, CYPB5M, Cytochrome b5 type B, OMB5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 66427, 80773, 80777 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43169, P04166, Q9CQX2 | |
| Cyb5b | |
| CYB5B Fusion Protein Ag7771 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |