missing translation for 'onlineSavingsMsg'
Learn More

CXCR6 Antibody (879112R) [Janelia Fluor™ 669], Novus Biologicals™

Código de producto. 30163628 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
Código de producto. Cantidad unitSize
30163628 0.1 mL 0.10 ml
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30163628

Marca: Novus Biologicals FAB8196RJF669

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCR6 Monoclonal antibody specifically detects CXCR6 in Rat samples. It is validated for Flow Cytometry,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antígeno CXCR6
Aplicaciones Flow Cytometry, CyTOF
Clasificación Monoclonal
Clon 879112R
Conjugado Janelia Fluor 669
Formulación 50mM Sodium Borate
Alias de gen BONZOG-protein coupled receptor bonzo, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33G-protein coupled receptor STRL33, TYMSTRCD186
Especie del huésped Mouse
Inmunógeno NS0 mouse myeloma cell line transfected with rat CXCR6
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites
Primario o secundario Primary
ID de gen (Entrez) 10663
Especies diana Rat
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.