missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
CXCR6 Monoclonal antibody specifically detects CXCR6 in Human samples. It is validated for Flow Cytometry, CyTOF
Especificaciones
Especificaciones
| Antígeno | CXCR6 |
| Aplicaciones | Flow Cytometry, CyTOF |
| Clasificación | Monoclonal |
| Clon | 1010910 |
| Conjugado | DyLight 350 |
| Dilución | Flow Cytometry, CyTOF-ready |
| Formulación | 50mM Sodium Borate |
| Alias de gen | BONZOG-protein coupled receptor bonzo, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33G-protein coupled receptor STRL33, TYMSTRCD186 |
| Especie del huésped | Mouse |
| Inmunógeno | HEK293 human embryonic kidney cell line transfected with human CXCR6, Met1-Leu342, Accession # O00574 |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?