Learn More
CXCR4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11073-2-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CXCR4 may be involved in WHIM syndrome (WHIMS). Immunodeficiency disease characterized by neutropenia, hypogammaglobulinemia and extensive human papillomavirus (HPV) infection. Despite the peripheral neutropenia, bone marrow aspirates from affected individuals contain abundant mature myeloid cells, a condition termed myelokathexis. The disease is caused by mutations affecting the gene represented in this entry.Especificaciones
| CXCR4 | |
| Polyclonal | |
| Unconjugated | |
| CXCR4 | |
| CD184, CXC R4, CXCR 4, CXCR4, CXCR4B, D2S201E, FB22, Fusin, HM89, HSY3RR, LAP3, LCR1, LESTR, SDF 1 receptor, WHIM | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12767, 60628, 7852 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O08565, P61073, P70658 | |
| Cxcr4 | |
| CXCR4 Fusion Protein Ag1528 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |