missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CXCR3 Polyclonal specifically detects CXCR3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Proximity Ligation Assay.
Specifications
Specifications
| Aplicaciones | Immunocytochemistry, Immunofluorescence |
| Clasificación | Polyclonal |
| Dilución | Western Blot 1 - 3 ug/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 31887215), Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen reported in scientific literature (PMID 19234157), Proximity Ligation Assay reported in scientific literature (PMID 34433033) |
| Alias de gen | CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR |
| Símbolos de los genes | CXCR3 |
| Método de purificación | Protein G purified |
| Contenido y almacenamiento | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Formulario | Purified |
| Isotype | IgG |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?