Learn More
CXCL8/IL8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Especificaciones
| Antígeno | CXCL8/IL8 |
|---|---|
| Concentración | 0.4 mg/mL |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|
16845385
|
Proteintech
27095-1-AP-20UL |
20 μL |
145.00€
20 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
|
16835385
|
Proteintech
27095-1-AP-150UL |
150 μL |
410.00€
150 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
Interleukin-8 (IL-8) is a chemokine produced by macrophages and other cell types such as epithelial cells. It is also synthesized by endothelial cells, which store IL-8 in their storage vesicles, the Weibel-Palade bodies. In humans, the interleukin-8 protein is encoded by the IL8 gene. Interleukin-8 (IL8) is a member of the CXC chemokine family. IL-8 is believed to play a role in the pathogenesis of bronchiolitis, a common respiratory tract disease caused by viral infection. This gene and other ten members of the CXC chemokine gene family form a chemokine gene cluster in a region mapped to chromosome 4q. The genes for IL8 have been co-localized on a 335-kb genomic fragment.Especificaciones
| CXCL8/IL8 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P10145 | |
| 3576 | |
| CXCL8/IL8 Fusion Protein Ag25640 | |
| Primary | |
| -20°C | |
| CXCL8 |
| 0.4 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| C X C motif chemokine 8, CXCL8, CXCL8,IL8, Emoctakin, GCP 1, GCP1, IL 8, IL 8(1 77), IL8, IL8/NAP1 form I Interleukin 8, IL8/NAP1 form II, IL8/NAP1 form VI, interleukin 8, MDNCF, MDNCF b IL 8(5 77) IL 8(6 77), NAF, NAF IL 8(7 77), NAP 1, NAP1, Neutrophil activating factor, Protein 3 10C | |
| CXCL8 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |