missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Interleukin-8 (IL-8) is a chemokine produced by macrophages and other cell types such as epithelial cells. It is also synthesized by endothelial cells, which store IL-8 in their storage vesicles, the Weibel-Palade bodies. In humans, the interleukin-8 protein is encoded by the IL8 gene. Interleukin-8 (IL8) is a member of the CXC chemokine family. IL-8 is believed to play a role in the pathogenesis of bronchiolitis, a common respiratory tract disease caused by viral infection. This gene and other ten members of the CXC chemokine gene family form a chemokine gene cluster in a region mapped to chromosome 4q. The genes for IL8 have been co-localized on a 335-kb genomic fragment.
Especificaciones
Especificaciones
| Antígeno | CXCL8/IL8 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.4 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | CXCL8 |
| N.º de referencia del gen | P10145 |
| Alias de gen | C X C motif chemokine 8, CXCL8, CXCL8,IL8, Emoctakin, GCP 1, GCP1, IL 8, IL 8(1 77), IL8, IL8/NAP1 form I Interleukin 8, IL8/NAP1 form II, IL8/NAP1 form VI, interleukin 8, MDNCF, MDNCF b IL 8(5 77) IL 8(6 77), NAF, NAF IL 8(7 77), NAP 1, NAP1, Neutrophil activating factor, Protein 3 10C |
| Símbolos de los genes | CXCL8 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?