missing translation for 'onlineSavingsMsg'
Learn More

CUGBP1/CELF1 Antibody (3B1), Biotin, Novus Biologicals™

Código de producto. 18233764 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
Código de producto. Cantidad unitSize
18233764 0.1 mL 0.10 ml
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18233764

Marca: Novus Biologicals NB200316B

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Denne vare kan ikke returneres. Se returpolitik

Mouse Monoclonal Antibody

CUGBP1/CELF1 Monoclonal specifically detects CUGBP1/CELF1 in Human, Mouse, Rat, Porcine, Bovine, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Tekniske data

Antígeno CUGBP1/CELF1
Aplicaciones Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Clasificación Monoclonal
Clon 3B1
Conjugado Biotin
Dilución Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulación PBS with 0.05% Sodium Azide
Alias de gen BRUNOL250 kDa nuclear polyadenylated RNA-binding protein, bruno-like 2, Bruno-like protein 2, CUG RNA-binding protein, CUG triplet repeat, RNA binding protein 1, CUG triplet repeat, RNA-binding protein 1, CUG-BP, CUG-BP- and ETR-3-like factor 1, CUGBP Elav-like family member 1, CUGBP, Elav-like family member 1, CUGBP1CUG triplet repeat RNA-binding protein 1, CUGBPCELF-1, Deadenylation factor CUG-BP, EDEN-BP, EDEN-BP homolog, embryo deadenylation element binding protein, Embryo deadenylation element-binding protein homolog, hNab50, NAB50CUG-BP1, NAPOR, nuclear polyadenylated RNA-binding protein, 50-kD, RNA-binding protein BRUNOL-2
Símbolos de los genes CELF1
Especie del huésped Mouse
Inmunógeno Full-length human CUGBP1 [UniProt# Q92879]
Método de purificación Protein G purified
Cantidad 0.1 mL
Disciplina de investigación Cancer, Chromatin Research, DNA replication Transcription Translation and Splicing, Neurodegeneration, Phospho Specific
Primario o secundario Primary
ID de gen (Entrez) 10658
Especies diana Human, Mouse, Rat, Pig, Bovine, Primate, Rabbit
Contenido y almacenamiento Store at 4C in the dark.
Formulario Purified
Isotype IgG1 κ
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.