missing translation for 'onlineSavingsMsg'
Learn More

Cubilin Antibody, Novus Biologicals™

Código de producto. 18645217 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
25 μL
Tamaño de la unidad:
100 microlitros
25 microlitros
Product Code. Cantidad unitSize
18645217 25 μL 25 microlitros
18608176 100 μg 100 microlitros
2 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 18645217

missing translation for 'mfr': Novus Biologicals NBP26261125ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cubilin Polyclonal antibody specifically detects Cubilin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Cubilin
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000
Formulación PBS (pH 7.2) and 40% Glycerol
Alias de gen cubilin, cubilin (intrinsic factor-cobalamin receptor), gp280, IFCR, MGA1
Especie del huésped Rabbit
Inmunógeno This antibody was developed against a recombinant protein corresponding to amino acids: SDQGFHITYLTSPSDLRCGGNYTDPEGELFLPELSGPFTHTRQCVYMMKQPQGEQIQINFTHVELQCQSDSSQNYIEVRDGETLLGKVCGNGTISHIKSITNSVWIRFKIDASVEKASFRAVYQVACGDELTGEGVIRSPFFPNVYPGE
Método de purificación Protein A purified
Cantidad 25 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 8029
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.