Learn More
CTGF/CCN2 Antibody (CL5339) [PerCP], Novus Biologicals™
Código de producto. 30288551 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30288551 0.1 mL 0.10 ml
1 options
Código de producto. 30288551
Marca: Novus Biologicals NBP261415PCP
Mouse Monoclonal Antibody
CTGF/CCN2 Monoclonal antibody specifically detects CTGF/CCN2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CTGF/CCN2
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon CL5339
Conjugado PerCP
Formulación PBS
Alias de gen CCN2IGFBP-8, connective tissue growth factor, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2
Especie del huésped Mouse
Inmunógeno This antibody was developed using a recombinant protein derived from P29279, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Angiogenesis, Cancer, Cell Biology, Cytokine Research, Growth and Development, Immunology, Innate Immunity
Primario o secundario Primary
ID de gen (Entrez) 1490
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1