missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CstF-64 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26825-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CSTF2 is one of three (including CSTF1 and CSTF3) cleavage stimulation factors which combine to form CSTF which is involved in the polyadenylation and 3′end cleavage of pre-mRNAs. CSTF2 contains a ribonucleoprotein-type RNA binding domain. CSTF2 is upregulated during activation of B cells which results in the switch of IgM heavy chain mRNA from membrane bound form to the secreted form.Especificaciones
| CstF-64 | |
| Polyclonal | |
| Unconjugated | |
| CSTF2 | |
| CF 1 64 kDa subunit, CstF 64, CSTF 64 kDa subunit, CSTF2, CstF-64 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 108062, 1478, 683927 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P33240, Q8BIQ5 | |
| Cstf2 | |
| CstF-64 Fusion Protein Ag25254 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido