missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
CSRP1 Polyclonal antibody specifically detects CSRP1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation, RNA immunoprecipitation
Especificaciones
Especificaciones
| Antígeno | CSRP1 |
| Aplicaciones | Western Blot, Immunoprecipitation |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:2000, Immunoprecipitation 1:50-1:200, RNA immunoprecipitation 1:20-1:50 |
| Formulación | PBS with 50% glycerol, pH7.3. |
| Alias de gen | CRP, CRP1, CSRPCYRP, cysteine and glycine-rich protein 1, Cysteine-rich protein 1, D1S181E, DKFZp686M148, LIM-domain protein |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant fusion protein containing a sequence corresponding to amino acids 1-193 of human CSRP1 (NP_004069.1). MPNWGGGKKCGVCQKTVYFAEEVQCEGNSFHKSCFLCMVCKKNLDSTTVAVHGEEIYCKSCYGKKYGPKGYGYGQGAGTLSTDKGESLGIKHEEAPGHRPTTNPNASKFAQKIGGSERCPRCSQAVYAAEKVIGAGKSWHKACFRCAKCGKGLESTTLADKDGEIYCKGCYAKNFGPKGFGFGQGAGALVHSE |
| Método de purificación | Affinity purified |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?