Learn More
Invitrogen™ Human CSNK1D (CK1 delta), GST Tag Recombinant Protein
Código de producto. 11563497
Change view
Click to view available options
Cantidad:
10 μg
Tamaño de la unidad:
10 microgramos
11563497 10 μg 10 microgramos
1 options
Código de producto. 11563497
Marca: Invitrogen™ PV3665
Recombinant Protein
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length.
CSNK1D (CK1 delta), a serine/threonine kinase, plays a critical role in Wnt/beta-catenin signaling pathway. This pathway is important in both development and cancer. The encoded protein may also be involved in the regulation of apoptosis, circadian rhythm, microtubule dynamics, chromosome segregation, and p53-mediated effects on growth. The encoded protein is highly similar to the mouse and rat CK1 delta homologs. Three transcript variants encoding different isoforms have been found for CK1 delta.
TRUSTED_SUSTAINABILITY
Especificaciones
Número de acceso P48730
Concentración See Label
Para utilizar con (aplicación) Kinase Assay
Formulación 50mM tris with 150mM NaCl, 50% glycerol, 0.02% Triton X-100, 2mM DTT, 0.5mM EDTA and no preservative; pH 7.5
ID de gen (Entrez) 1453
Peso molecular 74.3 kDa
Nombre Human CSNK1D (CK1 delta), GST Tag
Intervalo de pH 7.5
Método de purificación Purified
Cantidad 10 μg
Requisitos de almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Estado normativo RUO
Alias de gen 1200006A05Rik; AA409348; ASPS; casein kinase 1; casein kinase 1 delta; casein kinase 1, delta; casein kinase I isoform delta; CK1d; CK1delta; CKId; CKIdelta; CKI-delta; CSNK1D; D930010H05Rik; FASPS2; HCKID; HGNC:2452; Tau-protein kinase CSNK1D
Nombre común CK1 delta
Símbolo de gen CSNK1D
Familia Quinasa CK1 Kinase Family
Tipo de producto Protein
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína GST-tag
Secuencia Full length
Contenido y almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Entrada del ensayo Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity, Biochemical binding/kinetic
Grupo de cinasas Ser⁄Thr Kinases: CK1 Group
Condiciones de envío Dry Ice
Compatibilidad de sistemas LanthaScreen™ Antibody, Z'-LYTE™, Omnia™ Bulk Substrate
Entrada diana CSNK1D
Gen diana casein kinase 1, delta
Sistema de expresión Insect cells
Familia de proteínas Kinases & Inhibitors
Forma de proteína Recombinant, Full Length
Formulario Liquid
Subtipo de proteína Serine/Threonine Kinases
Categoría de investigación Oncology, Signal Transduction