Learn More
Abnova™ CSF1 (Human) Recombinant Protein
Código de producto. 16292290
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microgramos
50 microgramos
Código de producto. 16292290
Marca: Abnova™ P5818.100ug
Human CSF1 (159 a.a.) full-length recombinant protein with His tag expressed in Barley grain (Hordeum vulgare).
Especificaciones
Número de acceso BAD92189
Concentración 100 μg/mL
ID de gen (Entrez) 1435
Nombre colony stimulating factor 1 (macrophage)
Método de preparación Barley grain (Hordeum vulgare) expression system
Pruebas de control de calidad SDS-PAGE Stained with Coomassie Blue
Cantidad 100 μg
Requisitos de almacenamiento Store at -20°C on dry atmosphere. After reconstitution with sterile water to a concentration of no less than 100 ug/mL, store at -20°C. For long term storage it is recommended to add a carrier protein (0.1% HSA or BSA). Aliquot to avoid repeated freezing and thawing.
Alias de gen MCSF, MGC31930
Símbolo de gen CSF1
Especie Plants
Etiqueta de proteína His
Tampón Lyophilized from 5 mM TrisHCl, 80 mM Arginine, pH 8.0