missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CRTH-2/GPR44 Antibody (KHK patent anti-CRTH2) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Marca: Novus Biologicals™ NBP3-28373-50ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CRTH-2/GPR44 Monoclonal antibody specifically detects CRTH-2/GPR44 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Especificaciones
| CRTH-2/GPR44 | |
| Monoclonal | |
| Unconjugated | |
| Q9Y5Y4 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| KHK patent anti-CRTH2 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD294, CD294 antigen, Chemoattractant receptor-homologous molecule expressed on Th2 cells, CRTH2chemoattractant receptor homologous molecule expressed on T helper type 2 cells, DL1R, DP2, G protein-coupled receptor 44, putative G-protein coupled receptor 44 | |
| GPR44/ PTGDR2/ CD294 | |
| 50 μg | |
| GPCR, Innate Immunity | |
| 11251 | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido