Learn More
CRABP2 Mouse, anti-Human, CoraLite Plus 488, Clone: 1A5F3, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech CL488-66468-100UL
 Ver más versiones de este producto
Código de producto. 18048085
  • 450.00€ / 100 microlitros
Descripción
The CRABP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CRABP2. This antibody recognizes human, mouse, rat, pig antigen. The CRABP2 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Especificaciones
Especificaciones
CRABP2
1A5F3
CoraLite Plus 488
Mouse
Primary
Human, Mouse, Rat, Pig
155 kDa
IgG1
Flow Cytometry (Intracellular Staining)
1000 μg/mL
CRABP II, CRABP2, RBP6
100 μL
1382
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.