Learn More
CRABP2 Mouse, anti-Human, CoraLite Plus 488, Clone: 1A5F3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-66468-100UL
Descripción
The CRABP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CRABP2. This antibody recognizes human, mouse, rat, pig antigen. The CRABP2 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Especificaciones
| CRABP2 | |
| 1A5F3 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| 155 kDa | |
| IgG1 |
| Flow Cytometry (Intracellular Staining) | |
| 1000 μg/mL | |
| CRABP II, CRABP2, RBP6 | |
| 100 μL | |
| 1382 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.