missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COXIV Mouse anti-Human, Mouse, Clone: 3C7D2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60251-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is specific to COX4I1 and do not cross reacts with COX4I2.
Cytochrome c oxidase (COX) is the terminal enzyme of the mitochondrial respiratory chain. It is a multi-subunit enzyme complex that couples the transfer of electrons from cytochrome c to molecular oxygen and contributes to a proton electrochemical gradient across the inner mitochondrial membrane. The complex consists of 13 mitochondrial- and nuclear-encoded subunits. The functions of the nuclear-encoded subunits are unknown but they may play a role in the regulation and assembly of the complex. COX4 (COX4I1) is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.Especificaciones
| COXIV | |
| Monoclonal | |
| 1.07 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P13073, P19783 | |
| COX4I1 | |
| COXIV Fusion Protein Ag20551 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 3C7D2 | |
| Unconjugated | |
| COX4I1 | |
| COX IV 1, COX4, COX4I1, COXIV, Cytochrome c oxidase polypeptide IV, Cytochrome c oxidase subunit 4 isoform 1 mitochondrial | |
| Mouse | |
| Protein G | |
| RUO | |
| 12857, 1327 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido