Learn More
Fisherbrand™ Condenser Recycle Water and Heat
A standard condenser that collects water in a tank rather than letting water out into a drain.
en la oferta
Descripción
- Collects water in a tank
- Collected water can then be used for washing in the next cycle
- Heating element keeps temperature above 60°C to ensure that there is no possibility of bacteria developing in the tank
Especificaciones
Condensador |