missing translation for 'onlineSavingsMsg'
Learn More

COMMD1 Antibody (2B12), Novus Biologicals™

Product Code. 18590364 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mg
Unit Size:
0.10 miligramo
Product Code. Cantidad unitSize
18590364 0.1 mg 0.10 miligramo
1 options
This item is not returnable. View return policy

Product Code. 18590364

Brand: Novus Biologicals H00150684M03

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

COMMD1 Monoclonal antibody specifically detects COMMD1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antígeno COMMD1
Aplicaciones Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Sandwich ELISA
Clasificación Monoclonal
Clon 2B12
Conjugado Unconjugated
Formulación In 1x PBS, pH 7.4
N.º de referencia del gen AAH22046
Alias de gen C2orf5, COMM domain-containing protein 1, copper metabolism (Murr1) domain containing 1, copper metabolism gene MURR1, MGC27155, MURR1chromosome 2 open reading frame 5 (MURR1), Protein Murr1
Especie del huésped Mouse
Inmunógeno COMMD1 (AAH22046.1, 1 a.a. ∽ 190 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAAGELEGGKPLSGLLNALAQDTFHGYPGITEELLRSQLYPEVPPEEFRPFLAKMRGILKSIASADMDFNQLEAFLTAQTKKQGGITSDQAAVISKFWKSHKTKIRESLMNQSRWNSGLRGLSWRVDGKSQSRHSAQIHTPVAIIELELGKYGQESEFLCLEFDEVKVNQILKTLSEVEESISTLISQPN
Método de purificación IgG purified
Cantidad 0.1 mg
Estado normativo RUO
Disciplina de investigación Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 150684
Especies diana Human
Contenido y almacenamiento Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.