missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Collagen IV Monoclonal specifically detects Collagen IV in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.
Especificaciones
Especificaciones
| Aplicaciones | Immunohistochemistry (Frozen), Immunoblot |
| Símbolos de los genes | COL4A1 |
| Método de purificación | Affinity Purified |
| Cantidad | 0.1 mL |
| Estado normativo | RUO |
| Especies diana | Pig |
| Contenido y almacenamiento | Store at 4C in the dark. |
| Isotype | IgG1 |
For Research Use Only
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?