missing translation for 'onlineSavingsMsg'
Learn More

Collagen III alpha 1/COL3A1 Antibody (SR1186), Novus Biologicals™

Código de producto. p-200005365 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18627854 100 μg 100 microlitros
18667544 50 μg 50 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18627854 Proveedor Novus Biologicals N.º de proveedor NBP322424100ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Collagen III alpha 1/COL3A1 Monoclonal antibody specifically detects Collagen III alpha 1/COL3A1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Collagen III alpha 1/COL3A1
Aplicaciones Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation
Clasificación Monoclonal
Clon SR1186
Conjugado Unconjugated
Dilución Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50
Formulación PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Alias de gen alpha1 (III) collagen, alpha-1 type III collagen, COL3A1, Collagen 3, collagen alpha-1(III) chain, collagen, fetal, collagen, type III, alpha 1, Collagen-3, Collagen-III, collagenIII, EDS4A, EDSVASC, Ehlers-Danlos syndrome type IV, autosomal dominant, FLJ34534, PMGEDSV
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from Collagen III alpha 1/COL3A1 (Uniprot #: P02461)
Método de purificación Affinity purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Cancer, Cell Biology, Extracellular Matrix
Primario o secundario Primary
ID de gen (Entrez) 1281
Especies diana Human
Contenido y almacenamiento Store at -20° C. Avoid freeze/thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.