Learn More
CNPase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13427-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CNP may participate in RNA metabolism in the myelinating cell, CNP is the third most abundant protein in central nervous system myelin.Especificaciones
| CNPase | |
| Polyclonal | |
| Unconjugated | |
| CNP | |
| CNP, CNP1, CNPase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1267, 12799, 25275 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P09543, P13233, P16330 | |
| CNP | |
| CNPase Fusion Protein Ag4227 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |