missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNPase Antibody (1H10), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-50031
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CNPase Monoclonal antibody specifically detects CNPase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry Free-Floating
Especificaciones
| CNPase | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:5000 - 1:20000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:1000 - 1:5000, Immunohistochemistry Free-Floating | |
| P09543 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Free Floating) | |
| 1H10 | |
| Unconjugated | |
| 50% PBS, 50% glycerol | |
| 2', 3' cyclic nucleotide 3' phosphohydrolase, 2'-3'-cyclic nucleotide 3' phosphodiesterase, 2'-3'-cyclic-nucleotide 3'-phosphodiesterase, CNP1, CNPase, EC 3.1.4.37 | |
| Full length human recombinant CNPase protein expressed in and purified from E. coli. [UniProt# P09543] | |
| 0.1 mL | |
| Cellular Markers, Glia Markers, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers | |
| 1267 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido