missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
CML2 Polyclonal antibody specifically detects CML2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | CML2 |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Formulación | PBS with 50% glycerol, pH7.3. |
| Alias de gen | Camello-like protein 2, CML2, EC 2.3.1, EC 2.3.1.-, Hcml2, MGC97061, N-acetyltransferase 8B (GCN5-related, putative, gene/pseudogene), N-acetyltransferase 8B (gene/pseudogene), N-acetyltransferase Camello 2, NAT8BP, probable N-acetyltransferase 8B |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant fusion protein containing a sequence corresponding to amino acids 75-165 of human NAT8B (NP_057431.2). WFLAKKPWTRYVDIALRTDMSDITKSYLSECGSCFWVGESEEKVVGTVGALPVDDPTLREKRLQLFHLSVDNEHRGQGIAKALVRTVLQFA |
| Método de purificación | Affinity purified |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?