missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Clusterin/APOJ Antibody (AB-16B5) - IgG2SA, Novus Biologicals™

Código de producto. 30048468 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
Código de producto. Cantidad unitSize
30048468 50 μg 50 microgramos
30047089 100 μg 100 microgramos
30048813 1 mg 1 miligramo
3 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30048468

Marca: Novus Biologicals™ NBP32823350ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Human Monoclonal Antibody

Clusterin/APOJ Monoclonal antibody specifically detects Clusterin/APOJ in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Clusterin/APOJ
Aplicaciones Flow Cytometry, ELISA, Functional Assay
Clasificación Monoclonal
Clon AB-16B5
Conjugado Unconjugated
Formulación Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
N.º de referencia del gen Q96AJ1
Alias de gen 40, 40, sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLIclusterin (complement lysis inhibitor, SP-40, clusterin, Complement cytolysis inhibitor, complement lysis inhibitor, Complement-associated protein SP-40, Ku70-binding protein 1, KUB1SGP2, MGC24903, NA1/NA2, SGP-2, SP-40, sulfated glycoprotein 2, Testosterone-repressed prostate message 2, testosterone-repressed prostate message 2, apolipoprotein J), TRPM-2TRPM2
Especie del huésped Human
Inmunógeno Clusterin
Método de purificación Protein A purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Dendritic Cell Markers, Hypoxia, Immunology, Innate Immunity
Primario o secundario Primary
ID de gen (Entrez) 1191
Reconstitución Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Especies diana Human
Contenido y almacenamiento Store at -20°C in powder form. Store at -80°C once reconstituted.
Formulario Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.