Learn More
CLUH Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 30986-1-AP-20UL
Descripción
The CLUH antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CLUH. This antibody recognizes human, mouse antigen. The CLUH antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
| CLUH | |
| 150 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| CLU1, DKFZp686L05242, DKFZp686M09233, FLJ23672, KIAA0664, Protein KIAA0664 | |
| 20 μL | |
| 23277 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |