missing translation for 'onlineSavingsMsg'
Learn More

CLPP Antibody (OTI1F6), Allophycocyanin, Novus Biologicals™

Código de producto. 18184613 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18184613 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18184613 Proveedor Novus Biologicals N.º de proveedor NBP271363APC

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

CLPP Monoclonal specifically detects CLPP in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CLPP
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon OTI1F6
Conjugado APC
Dilución Flow Cytometry
Formulación PBS with 0.05% Sodium Azide
Alias de gen ATP-dependent protease ClpAP, proteolytic subunit, human, ClpP (caseinolytic protease, ATP-dependent, proteolytic subunit, E. coli)homolog, ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog (E.coli), ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog, ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog (E.coli), EC 3.4.21.92, Endopeptidase Clp, putative ATP-dependent Clp protease proteolytic subunit, mitochondrial
Símbolos de los genes CLPP
Especie del huésped Mouse
Inmunógeno Full length recombinant protein of human CLPP (NP_006003) produced in HEK293T cells.
Método de purificación Affinity Purified
Cantidad 0.1 mL
Disciplina de investigación Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 8192.0
Especies diana Human, Mouse, Rat, Canine, Monkey
Contenido y almacenamiento Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.