missing translation for 'onlineSavingsMsg'
Learn More

CLK1 Antibody - BSA Free, Novus Biologicals™

Código de producto. 18616972 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.02 mL
0.1 mL
Tamaño de la unidad:
0.01 ml
0.02 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18616972 0.02 mL 0.02 ml
18666422 0.1 mL 0.01 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 18616972 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP2924810.02ml

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CLK1 Polyclonal antibody specifically detects CLK1 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno CLK1
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1:500-1:2000
Formulación PBS with 50% glycerol, pH7.3.
Alias de gen CDC28/CDC2-like kinase, CDC-like kinase 1CLKEC 2.7.12.1, CLK/STY, dual specificity protein kinase CLK1, protein tyrosine kinase STY, STY
Especie del huésped Rabbit
Inmunógeno Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human CLK1 (NP_004062.2). MRHSKRTYCPDWDDKDWDYGKWRSSSSHKRRKRSHSSAQENKRCKYNHSKMCDSHYLESRSINEKDYHSRRYIDEYRNDYTQGCEPGHRQRDHESRYQNHSSKSSGRSGRSSYKSKHRIHHSTSHRRSHG
Método de purificación Affinity purified
Cantidad 0.02 mL
Estado normativo RUO
Disciplina de investigación Cell Cycle and Replication
Primario o secundario Primary
ID de gen (Entrez) 1195
Especies diana Human, Mouse
Contenido y almacenamiento Store at -20°C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.