missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC9a Antibody (14N8D7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-27087SS
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CLEC9a Monoclonal specifically detects CLEC9a in Human, Primate, Rhesus Macaque samples. It is validated for Western Blot.
Especificaciones
| CLEC9a | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1-3ug/ml | |
| C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for the antibody is 100% homologous in Human, Orangutan, Chimp and Rhesus Monkey, 80% in Dog, pand and Horse, 73% in Bovine and 60% homologous in Mouse. CLEC9A is a glycoprotein visualized by western blot between ∽25-45 kD. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot | |
| 14N8D7 | |
| Unconjugated | |
| Q6UXN8 | |
| CLEC9A | |
| A portion of amino acids 50-130 of human CLEC9A was used as the immunogen for the antibody. | |
| 0.025 mg | |
| Immunology, Stem Cells | |
| 283420 | |
| Human, Primate | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido