missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Citron Kinase Polyclonal specifically detects Citron Kinase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
| Antígeno | Citron Kinase |
| Aplicaciones | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulación | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| N.º de referencia del gen | O14578 |
| Alias de gen | CITK, citron (rho-interacting, serine/threonine kinase 21), Citron Rho-Interacting Kinase, Citron Rho-Interacting Serine/Threonine Kinase, CRIK, KIAA0949, MCPH17, Rho-interacting Serine/Threonine kinase 21, serine/threonine kinase 21, Serine/threonine-protein kinase 21, STK21 |
| Símbolos de los genes | CIT |
| Especie del huésped | Rabbit |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?