missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
cIAP-2/HIAP-1 Monoclonal antibody specifically detects cIAP-2/HIAP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
Specifications
| Antígeno | cIAP-2/HIAP-1 |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Clasificación | Monoclonal |
| Clon | SR2173 |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:50 |
| Formulación | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Alias de gen | AIP1, API2, BIRC3 baculoviral IAP repeat containing 3, c-IAP2, CIAP2, HAIP1, HIAP1, MALT2, MIHC, RNF49 |
| Especie del huésped | Rabbit |
| Inmunógeno | A synthesized peptide derived from human cIAP-2/HIAP-1 (Uniprot #: Q13489) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?