missing translation for 'onlineSavingsMsg'
Learn More

CHST12 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Código de producto. 18583872 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18583872 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Product Code. 18583872 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP297234AF700

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CHST12 Polyclonal antibody specifically detects CHST12 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno CHST12
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Alexa Fluor 700
Formulación 50 mM Sodium Borate
Alias de gen C4S-2, C4ST2C4ST-2, carbohydrate (chondroitin 4) sulfotransferase 12, carbohydrate sulfotransferase 12, Chondroitin 4-O-sulfotransferase 2, Chondroitin 4-sulfotransferase 2, EC 2.8.2.5, Sulfotransferase Hlo
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with E. coli-derived Human CHST12 fragment.
Método de purificación Antigen and protein A Affinity-purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cancer, Cell Biology, Cytoskeleton Markers, Endocrinology, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 55501
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.