missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHN2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18259-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene is a member of the chimerin family and encodes a protein with a phorbol-ester/DAG-type zinc finger, a Rho-GAP domain and an SH2 domain. This protein has GTPase-activating protein activity that is regulated by phospholipid binding and binding of diacylglycerol (DAG) induces translocation of the protein from the cytosol to the Golgi apparatus membrane. The protein plays a role in the proliferation and migration of smooth muscle cells. Decreased expression of this gene is associated with high-grade gliomas and breast tumors, and increased expression of this gene is associated with lymphomas. Mutations in this gene have been associated with schizophrenia in men. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Especificaciones
| CHN2 | |
| Polyclonal | |
| Unconjugated | |
| CHN2 | |
| ARHGAP3, Beta chimaerin, Beta chimerin, chimerin (chimaerin) 2, CHN2, RHOGAP3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1124, 69993 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P52757, Q80XD1 | |
| Chn2 | |
| CHN2 Fusion Protein Ag13106 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido