Learn More
CHCHD10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25671-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CHCHD10 encodes a mitochondrial protein that is enriched at cristae junctions in the intermembrane space. It may play a role in cristae morphology maintece or oxidative phosphorylation. Mutations in this gene cause frontotemporal dementia and/or amyotrophic lateral sclerosis-2. Alternative splicing of this gene results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 7 and 19.Especificaciones
| CHCHD10 | |
| Polyclonal | |
| Unconjugated | |
| CHCHD10 | |
| C22orf16, CHCHD10, N27C7 4, Protein N27C7 4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 103172, 400916 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8WYQ3 | |
| CHCHD10 | |
| CHCHD10 Fusion Protein Ag22598 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |