missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGGBP1 Mouse anti-Human, Rat, Clone: 2F5G9, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66524-1-IG-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a CGG repeat-binding protein that primarily localizes to the nucleus. CGG trinucleotide repeats are implicated in many disorders as they often act as transcription- and translation-regulatory elements, can produce hairpin structures which cause DNA replication errors, and form regions prone to chromosomal breakage. CGG repeats are also targets for CpG methylation. In addition to its ability to bind CGG repeats and regulate transcription, this gene is believed to play a role in DNA damage repair and telomere protection. In vitro studies indicate this protein does not bind to methylated CpG sequences.Especificaciones
| CGGBP1 | |
| Monoclonal | |
| 0.93 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UFW8 | |
| Cggbp1 | |
| CGGBP1 Fusion Protein Ag1129 | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 2F5G9 | |
| Unconjugated | |
| CGGBP1 | |
| 20 kDa CGG binding protein, CGG binding protein 1, CGGBP, CGGBP1, p20 CGGBP, p20 CGGBP DNA binding protein | |
| Mouse | |
| Protein G | |
| RUO | |
| 288353, 8545 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido