Learn More
CFD Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26050-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Complement factor D is a serine protease of the alternative pathway of complement activation. The protein is a single chain polypeptide with a molecular mass of 25 kDa. Factor D cleaves factor B bound to C3b, generating the alternative pathway C3 convertase C3bBb and releasing the Ba fragment.Especificaciones
| CFD | |
| Polyclonal | |
| Unconjugated | |
| CFD | |
| ADIPSIN, ADN, C3 convertase activator, CFD, Complement factor D, complement factor D (adipsin), DF, PFD, Properdin factor D | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1675 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P00746 | |
| CFD | |
| CFD Fusion Protein Ag23473 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |