missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CEND1 Monoclonal antibody specifically detects CEND1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
Specifications
| Antígeno | CEND1 |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Clasificación | Monoclonal |
| Clon | SR1342 |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Formulación | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Alias de gen | BM88 antigen, cell cycle exit and neuronal differentiation 1, cell cycle exit and neuronal differentiation protein 1, FLJ90066 |
| Especie del huésped | Rabbit |
| Inmunógeno | A synthesized peptide derived from human CEND1 (Uniprot #: Q8N111) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?