missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM8 Recombinant Rabbit Monoclonal Antibody (013), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
256.00€ - 510.00€
Especificaciones
Antígeno | CEACAM8 |
---|---|
Clon | 013 |
Concentración | 1 mg/mL |
Aplicaciones | ELISA |
Clasificación | Recombinant Monoclonal |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
17799803
|
Invitrogen
MA543461 |
50 μL |
256.00€
50 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
17709633
|
Invitrogen
MA543076 |
100 μL |
510.00€
100 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
Specificity: Human CEACAM8/CD66b
This gene encodes a member of the carcinoembryonic antigen (CEA) gene family, which belongs to the immunoglobulin superfamily. Two subgroups of the CEA family, the CEA cell adhesion molecules and the pregnancy-specific glycoproteins, are located within a 1.2 Mb cluster on the long arm of chromosome 19. Eleven pseudogenes of the CEA cell adhesion molecule subgroup are also found in the cluster. The encoded protein was originally described in bile ducts of liver as biliary glycoprotein. Subsequently, it was found to be a cell-cell adhesion molecule detected on leukocytes, epithelia, and endothelia. The encoded protein mediates cell adhesion via homophilic as well as heterophilic binding to other proteins of the subgroup. Multiple cellular activities have been attributed to the encoded protein, including roles in the differentiation and arrangement of tissue three-dimensional structure, angiogenesis, apoptosis, tumor suppression, metastasis, and the modulation of innate and adaptive immune responses. Multiple transcript variants encoding different isoforms have been reported, but the full-length nature of all variants has not been defined.Especificaciones
CEACAM8 | |
1 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
carcinoembryonic antigen CGM6; carcinoembryonic antigen gene family member 6; carcinoembryonic antigen related cell adhesion molecule 8; carcinoembryonic antigen-related cell adhesion molecule 8; CD66b; CD67; CD67 antigen; CEACAM8; CGM6; NCA-95; non-specific cross-reacting antigen NCA-95 | |
CEACAM8 | |
IgG | |
Protein A | |
Antibody |
013 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
P31997 | |
1088 | |
Recombinant Human CEACAM8/CD66b protein. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
CEACAM8 |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
CEACAM8 Recombinant Rabbit Monoclonal Antibody (013), Invitrogen™