missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM6/CD66c Antibody (439424), mFluor Violet 610 SE, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
CEACAM6/CD66c Monoclonal antibody specifically detects CEACAM6/CD66c in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Especificaciones
Especificaciones
| Antígeno | CEACAM6/CD66c |
| Aplicaciones | Western Blot, Flow Cytometry, CyTOF |
| Clasificación | Monoclonal |
| Clon | 439424 |
| Conjugado | mFluor Violet 610 SE |
| Dilución | Western Blot, Flow Cytometry, CyTOF-ready |
| Formulación | 50mM Sodium Borate |
| Alias de gen | carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen |
| Especie del huésped | Mouse |
| Inmunógeno | Mouse myeloma cell line NS0-derived recombinant human CEACAM-6/CD66c, Lys35-Gly320, Accession # P40199 |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?