missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CDK6 (Human) Recombinant Protein
Human CDK6 full-length ORF ( NP_001250.1, 1 a.a. - 326 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00001021-P02.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MEKDGLCRADQQYECVAEIGEGAYGKVFKARDLKNGGRFVALKRVRVQTGEEGMPLSTIREVAVLRHLETFEHPNVVRLFDVCTVSRTDRETKLTLVFEHVDQDLTTYLDKVPEPGVPTETIKDMMFQLLRGLDFLHSHRVVHRDLKPQNILVTSSGQIKLADFGLARIYSFQMALTSVVVTLWYRAPEVLLQSSYATPVDLWSVGCIFAEMFRRKPLFRGSSDVDQLGKILDVIGLPGEEDWPRDVALPRQAFHSKSAQPIEKFVTDIDELGKDLLLKCLTFNPAKRISAYSALSHPYFQDLERCKENLDSHLPPSQNTSELNTA
Especificaciones
NP_001250.1 | |
cyclin-dependent kinase 6 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDK6 | |
GST |
1021 | |
Wheat germ expression system | |
10 μg | |
MGC59692, PLSTIRE, STQTL11 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |