missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD99 Monoclonal antibody specifically detects CD99 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antígeno | CD99 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | MIC2/8119R |
| Concentración | 0.2 mg/ml |
| Conjugado | Unconjugated |
| Dilución | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Formulación | 10mM PBS with 0.05% BSA |
| Alias de gen | 12E7, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2 |
| Especie del huésped | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?